Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

033201-A1
AA33201-A1EA 85.5 USD
AA33201-A1
Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)
Aluminium oxide

Contains 0.5% organic binder to increase dry strength


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: Agglomerates 150 micron Odor: Odorless MDL No.: MFCD00211795

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00211795
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR