Combustion Trays, Aluminium Oxide AL24, Thermo Scientific
Supplier: Thermo Scientific Chemicals
033187-KF
AA33187-KFEA 405.9 USD
AA33187-KF
Combustion Trays, Aluminium Oxide AL24, Thermo Scientific
Boats Combustion Boats
Pure Al2O3 Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.