Cyanogen bromide
Supplier: Thermo Scientific Chemicals
|
Danger
|
032731-22
032731-06
032731-14
AA32731-22EA
158
USD
AA32731-22
AA32731-06
AA32731-14
Cyanogen bromide
Cyanogen bromide
MDL: MFCD00011597
CAS No.: 506-68-3
Molecular Formula: BrCN
Formula Weight: 105.93
Physical Form: Crystalline
Density: 2.015
Odor: Acrid
Boiling Point: 61.4°
Merck Reference: 14,2693
UN No.: 1889
MDL No.: MFCD00011597
Formula:
CBrN Boiling Pt: 61.4 °C Melting Pt: 50…52 °C Density: 2.015 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011597 CAS Number: 506-68-3 EINECS: 208-051-2 UN: 1889 ADR: 6.1,I Merck Index: 14,2693 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...