Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼ 2mm), with indicator

Supplier: Thermo Scientific Chemicals
Danger

032256-30 032256-A1 032256-A7
AA32256-30EA 84.09 USD
AA32256-30 AA32256-A1 AA32256-A7
Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼ 2mm), with indicator
Molecular sieve A4 (0.4 nm, 4 Å)
Storage Temperature: Ambient
MDL Number: MFCD00147627
CAS Number: 70955-01-0
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR