Arsenic(III) chloride ≥99%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00014165
Decomposes in water with formation of As(OH)3 and HCl. Increasing dilutions in water result in As2O3 precipitation
CAS No.: 7784-34-1
Molecular Formula: AsCl3
Formula Weight: 181.27
Physical Form: Liquid
Density: 2.163
Sensitivity: Moisture Sensitive
Boiling Point: 130.2°
Solubility: Decomposes in water with formation of As(OH)3 and HCl. Increasing dilutions in water result in As2O3 precipitation
Merck Reference: 14,801
UN No.: 1560
MDL No.: MFCD00014165
Formula:
AsCl₃ Boiling Pt: 130.2 °C Melting Pt: –16 °C Density: 2.163 Storage Temperature: Ambient |
MDL Number:
MFCD00014165 CAS Number: 7784-34-1 EINECS: 232-059-5 UN: 1560 ADR: 6.1,I Merck Index: 14,801 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...