1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4,8,11-Tetramethylcyclam

030854-03
AA30854-03EA 292.15 USD
AA30854-03
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

CAS No.: 41203-22-9 Molecular Formula: C14H32N4 Formula Weight: 256.43 Flash Point: 110°(230°F) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 34-36° MDL No.: MFCD00005106

Formula: C₁₄H₃₂N₄
Melting Pt: 34…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005106
CAS Number: 41203-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR