Tetramethylammonium hydroxide 25% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

030833-36 030833-09 030833-22
AA30833-36EA 141.03 USD
AA30833-36 AA30833-09 AA30833-22
Tetramethylammonium hydroxide 25% (w/w) in methanol
Tetramethylammonium hydroxide

MDL: MFCD00008280


CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Flash Point: 12°(54°F) Physical Form: Liquid Density: 0.865 Appearance: Colorless to pale yellow Odor: Like alcohol Merck Reference: 14,9224 UN No.: 2924 MDL No.: MFCD00008280

Formula: C₄H₁₃NO
MW: 91.15
Density: 0.865
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 2924
ADR: 3 II
Merck Index: 14,09224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR