Lithium tri-tert-butoxyaluminum hydride ≥94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithium tri-tert-butoxyaluminohydride
030716-09
AA30716-09EA
104
USD
AA30716-09
Lithium tri-tert-butoxyaluminum hydride ≥94%
Lithium tri-tert-butoxyaluminum hydride
Purity: 94%
CAS No.: 17476-04-9
Molecular Formula: C12H28AlLiO3
Formula Weight: 254.28
Physical Form: Powder
Density: 1.03
Appearance: White to pale grey
Odor: Odorless
Melting Point: ca 319° dec.
Beilstein Reference: 5796791
UN No.: 1409
MDL No.: MFCD00011532
Formula:
C₁₂H₂₈AlLiO₃ Melting Pt: ∼ 319 °C (dec.) Density: 1,03 Storage Temperature: Ambient |
MDL Number:
MFCD00011532 CAS Number: 17476-04-9 EINECS: 241-490-8 UN: 1409 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...