2,2'-Bipyridyl 98% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
030569-18
030569-30
030569-09
AA30569-18EA
104.96
USD
AA30569-18
AA30569-30
AA30569-09
2,2'-Bipyridyl 98% ACS
2,2'-Bipyridyl
MDL: MFCD00006212
Beilstein Registry No.: 113089
Purity: 98%
CAS No.: 366-18-7
Molecular Formula: C10H8N2
Formula Weight: 156.19
Flash Point: 121°(249°F)
Physical Form: Crystalline
Melting Point: 70°
Boiling Point: 272-273°
Beilstein Reference: 113089
Merck Reference: 14,3347
UN No.: 2811
MDL No.: MFCD00006212
Formula:
C₁₀H₈N₂ Boiling Pt: 272…273 °C Melting Pt: 70 °C Density: 1.326 g/cm³ (–150 °C) Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III Merck Index: 14,3347 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...