Strontium nitrate, anhydrous ≥99.97% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

030486-14 030486-22
AA30486-14EA 145 USD
AA30486-14 AA30486-22
Strontium nitrate, anhydrous ≥99.97% (metals basis)
Strontium nitrate

MDL: MFCD00011248


Purity: 99.97% CAS No.: 10042-76-9 Molecular Formula: N2O6Sr Formula Weight: 211.63 Physical Form: Crystalline Density: 2.990 Appearance: White Odor: Odorless Melting Point: 570° Merck Reference: 14,8845 UN No.: 1507 MDL No.: MFCD00011248

Formula: Sr(NO₃)₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.990
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 14,8845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR