Potassium sulfate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
030485-06
030485-14
030485-22
AA30485-06EA
51.7
USD
AA30485-06
AA30485-14
AA30485-22
Potassium sulfate ≥99.99% (metals basis)
Potassium sulphate
MDL: MFCD00011388
Purity: 99.99%
CAS No.: 7778-80-5
Molecular Formula: K2SO4
Formula Weight: 174.27
Physical Form: Powder
Density: 2.662
Melting Point: 1069°
Boiling Point: 1689°
Merck Reference: 14,7674
MDL No.: MFCD00011388
Formula:
K₂SO₄ MW: 174.26 g/mol Boiling Pt: 1698 Melting Pt: 1069 Density: 2.662 Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 14,7674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...