Trimethylphosphine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl phosphine

ampouled under argon

030143-06 030143-14 030143-03
AA30143-06EA 203.37 USD
AA30143-06 AA30143-14 AA30143-03
Trimethylphosphine 98%
Trimethylphosphine

It is used in the preparation of organic phosphorus compounds and frequently-used phosphine ligand. It is also a highly basic ligand that forms complexes with most metals. In addition, it can react with bromomethyl-benzene to get benzyl-trimethyl-phosphonium; bromide.


Purity: 99% CAS No.: 594-09-2 Molecular Formula: (CH3)3P Formula Weight: 76.08 Flash Point: -30°(-22°F) Physical Form: Liquid, Ampouled under argon Density: 0.735 Sensitivity: Air and Moisture Sensitive Odor: Pungent Boiling Point: 38-40° UN No.: 2845 MDL No.: MFCD00008510

Formula: C₃H₉P
Boiling Pt: 38…40 °C
Melting Pt: –77.8 °C
Density: 0.735
Flash Pt: –30 °C (–22 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR