Dicarbonylcyclopentadienylcobalt
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)
023136-03
023136-06
AA23136-03EA
51.6
USD
AA23136-03
AA23136-06
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt
Applications: Catalyst for the cyclotrimerization of alkynes
CAS No.: 12078-25-0
Molecular Formula: C7H5CoO2
Formula Weight: 180.05
Flash Point: 26°(80°F)
Physical Form: Liquid
Sensitivity: Air Sensitive
Boiling Point: 139-140°/710mm
UN No.: 3281
MDL No.: MFCD00009589
Applications: Catalyst for the cyclotrimerization of alkynes
Formula:
C₇H₅CoO₂ Boiling Pt: 139…140 °C (710 mmHg) Flash Pt: 26 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009589 CAS Number: 12078-25-0 EINECS: 235-139-8 UN: 3281 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...