Trimethyl borate ≥99.9995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

022975-30 022975-18
AA22975-30EA 271 USD
AA22975-30 AA22975-18
Trimethyl borate ≥99.9995% (metals basis)
Trimethyl borate

MDL: MFCD00008346 Beilstein Registry No.: 1697939 Fieser: 1,673


Purity: 99.9995+% CAS No.: 121-43-7 Molecular Formula: C3H9BO3 Formula Weight: 103.92 Flash Point: -8°(17°F) Physical Form: Liquid Density: 0.915 Refractive Index: 1.3610 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Odorless Melting Point: -34° Boiling Point: 67-69° Beilstein Reference: 1697939 Merck Reference: 14,9712 UN No.: 2416 MDL No.: MFCD00008346

Formula: C₃H₉BO₃
Boiling Pt: 67…69 °C
Melting Pt: –34 °C
Density: 0.915
Flash Pt: –8 °C (17 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II
Merck Index: 14,9712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR