Specpure® Sulfur, AAS Standard Solution, S 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
022958-AE
022958-AP
AA22958-AEEA
40.77
USD
AA22958-AE
AA22958-AP
Specpure® Sulfur, AAS Standard Solution, S 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Sulfur standards
Molecular Formula: (NH4)2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00085316
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...