Vanadium(IV) oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Vanadium dioxide
022957-18
022957-09
AA22957-18EA
367
USD
AA22957-18
AA22957-09
Vanadium(IV) oxide ≥99% (metals basis)
Vanadium (IV) oxide
MDL: MFCD00049702
Purity: 99%
CAS No.: 12036-21-4
Molecular Formula: O2V, may contain V6O13
Formula Weight: 82.94
Physical Form: -100 Mesh Powder
Density: 4.339
Appearance: Black
Melting Point: 1967°
UN No.: 3285
MDL No.: MFCD00049702
Formula:
O₂V Boiling Pt: 2700 °C (1013 hPa) Melting Pt: 1967 °C Density: 4,339 Storage Temperature: Ambient |
MDL Number:
MFCD00049702 CAS Number: 12036-21-4 EINECS: 234-841-1 UN: 3285 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...