Acetonitrile, dehydrated (max. 0.01% H₂O) ≥99.7% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

022927-K2
AA22927-K2EA 133 USD
AA22927-K2 AA22927-M6 AA22927-M1 AAA22927-K7
Acetonitrile, dehydrated (max. 0.01% H₂O) ≥99.7% for HPLC
Acetonitrile

Acetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.


Filtered through 0.2 µ filters. Incompatible with alkali metals, acids, bases, reducing agents and oxidizing agents. Highly flammable.

Formula: H₃CCN
Boiling Pt: 80…82 °C
Melting Pt: –48… –45 °C
Density: 0.786
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
Merck Index: 14,70
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99.7%
Residual Water 0.01%
Pesticide Residue Analysis Passes test
Residue on Evaporation 0.0001%
UV Transmittance: 190 nm ≥10%
UV Transmittance: 195 nm ≥50%
UV Transmittance: 200 nm ≥80%
UV Transmittance: 225 nm ≥95%
UV Transmittance: 240-400 nm ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR