Iron(II) sulfide ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ferrous sulphide, Ferrous sulfide
022388-06
022388-14
AA22388-06EA
98.2
USD
AA22388-06
AA22388-14
Iron(II) sulfide ≥99.98% (metals basis)
Iron (II) sulphide
MDL: MFCD00011013
Purity: 99.98%
CAS No.: 1317-37-9
Molecular Formula: FeS
Formula Weight: 87.91
Physical Form: Powder
Density: 4.74
Appearance: Brown to black
Odor: Odorless
Melting Point: 1195°
Merck Reference: 14,4058
MDL No.: MFCD00011013
Formula:
FeS Melting Pt: 1195 °C Density: 4.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 UN: 3077 ADR: 9,III Merck Index: 14,4058 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...