1,2-Dichlorobenzene ≥98% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
o-Dichlorobenzene
019385-M1
019385-K2
019385-K7
AA19385-M1EA
365.04
USD
AA19385-M1
AA19385-K2
AA19385-K7
1,2-Dichlorobenzene ≥98% for HPLC
1,2-Dichlorobenzene
MDL: MFCD00000535
Beilstein Registry No.: 606078
Notes: Filtered through 0.2µ filters.
Purity: 98% min
CAS No.: 95-50-1
Molecular Formula: C6H4Cl2
Formula Weight: 147.00
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.306
Refractive Index: 1.5510
Odor: Pungent
Melting Point: -18°
Boiling Point: 180°
Beilstein Reference: 606078
Merck Reference: 14,3056
UN No.: 1591
MDL No.: MFCD00000535
Formula:
C₆H₄Cl₂ MW: 147 g/mol Boiling Pt: 180.5 Melting Pt: -17 Density: 1.306 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000535 CAS Number: 95-50-1 EINECS: 202-425-9 UN: 1591 ADR: 6.1 III Merck Index: 14,3056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...