1,2-Dichlorobenzene ≥98% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00000535
Beilstein Registry No.: 606078
Notes: Filtered through 0.2µ filters.
Purity: 98% min
CAS No.: 95-50-1
Molecular Formula: C6H4Cl2
Formula Weight: 147.00
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.306
Refractive Index: 1.5510
Odor: Pungent
Melting Point: -18°
Boiling Point: 180°
Beilstein Reference: 606078
Merck Reference: 14,3056
UN No.: 1591
MDL No.: MFCD00000535
Formula:
C₆H₄Cl₂ Boiling Pt: 180 °C Melting Pt: –18 °C Density: 1.306 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000535 CAS Number: 95-50-1 EINECS: 202-425-9 UN: 1591 ADR: 6.1,III Merck Index: 14,3056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...