Tetracyclohexyltin

Supplier: Thermo Scientific Chemicals
Danger

018822-06 018822-14 018822-03
AA18822-06EA 342 USD
AA18822-06 AA18822-14 AA18822-03
Tetracyclohexyltin
Tetracyclohexyltin

MDL: MFCD00014278


CAS No.: 1449-55-4 Molecular Formula: C24H44Sn Formula Weight: 451.31 Physical Form: Powder Melting Point: 241-250° UN No.: 3146 MDL No.: MFCD00014278

Formula: C₂₄H₄₄Sn
Melting Pt: 241…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00014278
CAS Number: 1449-55-4
EINECS: 215-910-5
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR