1-Bromopyrene ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
018280-03
018280-06
AA18280-03EA
54
USD
AA18280-03
AA18280-06
1-Bromopyrene ≥95%
1-Bromopyrene
MDL: MFCD00015767
Purity: 95%
CAS No.: 1714-29-0
Molecular Formula: C16H9Br
Formula Weight: 281.13
Physical Form: Crystalline
Melting Point: 94-95°
MDL No.: MFCD00015767
Formula:
C₁₆H₉Br Melting Pt: 94…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015767 CAS Number: 1714-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...