Bis(2-ethylhexyl)phosphate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
017723-22
017723-36
017723-A3
AA17723-22EA
40.39
USD
AA17723-22
AA17723-36
AA17723-A3
Bis(2-ethylhexyl)phosphate ≥95%
Bis(2-ethylhexyl) phosphate
Applications: Extraction of metals
Purity: 95%
CAS No.: 298-07-7
Molecular Formula: C16H35O4P
Formula Weight: 322.43
Flash Point: 171°(340°F)
Physical Form: Liquid
Density: 0.965
Refractive Index: 1.4420
Appearance: Colorless
pH: 2-3
Melting Point: -50°
Boiling Point: 48°/12mm
UN No.: 1902
MDL No.: MFCD00009492
Extraction of metals
Formula:
C₁₆H₃₅O₄P Boiling Pt: 48 °C (12 mmHg) Melting Pt: –50 °C Density: 0.965 Flash Pt: 171 °C (340 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009492 CAS Number: 298-07-7 EINECS: 206-056-4 UN: 1902 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...