Sodium orthosilicate (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetrasodium orthosilicate, Sodium silicate (ortho)

017568-A1
AA17568-A1EA 61.88 USD
AA17568-A1
Sodium orthosilicate (Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)
tetra-Sodium orthosilicate

It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.


CAS No.: 13472-30-5 Molecular Formula: Na4SiO4 Physical Form: Crystalline Appearance: White Odor: Odorless UN No.: 1759 MDL No.: MFCD00014252

Formula: Na₄O₄Si
MW: 184.04 g/mol
Melting Pt: 1078 °C
Density: 2,5 g/cm³ (26 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014252
CAS Number: 13472-30-5
EINECS: 236-741-3
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR