Iron(II) sulfide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ferrous sulphide, Ferrous sulfide
017422-18
017422-09
AA17422-18EA
202
USD
AA17422-18
AA17422-09
Iron(II) sulfide ≥99% (metals basis)
Iron (II) sulphide
MDL: MFCD00011013
Soluble in acids with H2S evolution
Purity: 99%
CAS No.: 1317-37-9
Molecular Formula: FeS (Fe typically 0.9)
Formula Weight: 87.91
Physical Form: -100 Mesh Powder
Density: 4.74
Appearance: Brown to black
Odor: Odorless
Melting Point: 1195°
Solubility: Soluble in acids with H2S evolution
Merck Reference: 14,4058
MDL No.: MFCD00011013
Formula:
FeS Melting Pt: 1195 °C Density: 4.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 UN: 3077 ADR: 9,III Merck Index: 14,4058 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...