Strontium chloride, anhydrous ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

016790-14 016790-22 016790-36
AA16790-14EA 108 USD
AA16790-14 AA16790-22 AA16790-36
Strontium chloride, anhydrous ≥99.5% (metals basis)
Strontium chloride

MDL: MFCD00011249


Purity: 99.5% CAS No.: 10476-85-4 Molecular Formula: Cl2Sr Formula Weight: 158.53 Physical Form: -40 Mesh Powder Density: 3.052 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 873° Boiling Point: 1250° Merck Reference: 14,8840 MDL No.: MFCD00011249

Formula: Cl₂Sr
MW: 158.53
Boiling Pt: 1250
Melting Pt: 868
Density: 3.052
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 14,8840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR