Trimethylolpropane triacrylate stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
016445-22
016445-A3
016445-36
AA16445-22EA
60.8
USD
AA16445-22
AA16445-A3
AA16445-36
Trimethylolpropane triacrylate stabilized
Trimethylolpropane triacrylate
CAS No.: 15625-89-5
Molecular Formula: (CH2=CHCO2CH2)3CCH2CH3
Formula Weight: 296.31
Flash Point: 87°(188°F)
Physical Form: Liquid
Density: 1.10
Refractive Index: 1.4735
Odor: Pungent
Boiling Point:
Formula:
C₁₅H₂₀O₆ Boiling Pt: >200 °C (1 mmHg) Density: 1,10 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008628 CAS Number: 15625-89-5 EINECS: 239-701-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...