Phenol red ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

016294-06 016294-14 016294-22
AA16294-06EA 48.75 USD
AA16294-06 AA16294-14 AA16294-22
Phenol red ACS
Phenol red

pH indicator


CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Physical Form: Crystalline Odor: Odorless Melting Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552


pH indicator

Formula: C₁₉H₁₄O₅S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 14,7247

Order Now

Specification Test Results

Clarity of solution Pass test
Visual transition interval pH 6.8 (yellow) to 8.2 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR