1-Pentanesulfonic acid sodium salt monohydrate for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate

015714-06 015714-22 015714-14
AA15714-06EA 54.8 USD
AA15714-06 AA15714-22 AA15714-14
1-Pentanesulfonic acid sodium salt monohydrate for HPLC
1-Pentanesulphonic acid sodium salt monohydrate

Ion pair reagent


CAS No.: 207605-40-1 Molecular Formula: C5H11NaO3S.H2O Formula Weight: 192.21 (174.19anhy) Physical Form: Crystalline powder Appearance: White Beilstein Reference: 3725147


Ion pair reagent

Formula: C₅H₁₁NaO₃S·1H₂O
MW: 192.21
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 207605-40-1
EINECS: 245-208-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR