1-Pentanesulfonic acid sodium salt monohydrate for HPLC
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate
015714-06
015714-22
015714-14
AA15714-06EA
54.8
USD
AA15714-06
AA15714-22
AA15714-14
1-Pentanesulfonic acid sodium salt monohydrate for HPLC
1-Pentanesulphonic acid sodium salt monohydrate
Ion pair reagent
CAS No.: 207605-40-1
Molecular Formula: C5H11NaO3S.H2O
Formula Weight: 192.21 (174.19anhy)
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 3725147
Ion pair reagent
Formula:
C₅H₁₁NaO₃S·1H₂O MW: 192.21 Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 207605-40-1 EINECS: 245-208-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...