Copper(II) gluconate
Supplier: Thermo Scientific Chemicals
Synonyms:
Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
015325-18
015325-A1
015325-30
AA15325-18EA
32.4
USD
AA15325-18
AA15325-A1
AA15325-30
Copper(II) gluconate
Copper(II) gluconate
CAS No.: 527-09-3
Molecular Formula: [HOCH2[CH(OH)]4CO2]2Cu
Formula Weight: 453.85
Physical Form: Powder
Melting Point: 155-157°
MDL No.: MFCD00075297
Formula:
C₆H₁₂O₇ Melting Pt: 155…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075297 CAS Number: 527-09-3 EINECS: 208-408-2 Merck Index: 14,2640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...