Gold ≥99.95% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
014729-BQ
014729-BU
014729-BO
AA14729-BQEA
868
USD
AA14729-BQ
AA14729-BU
AA14729-BO
Gold ≥99.95% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Gold
MDL: MFCD00003436
Purity: 99.95%
CAS No.: 7440-57-5
Physical Form: ≈60.6g/m
Odor: Odorless
MDL No.: MFCD00003436
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...