Gold ≥99.999% (metals basis), wire, Premion®, Diameter 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

014725-BW 014725-CB 014725-BS
AA14725-BWEA 544 USD
AA14725-BW AA14725-CB AA14725-BS
Gold ≥99.999% (metals basis), wire, Premion®, Diameter 0.5 mm (0.02 in)
Gold

MDL: MFCD00003436


Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈3.79g/m Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR