Niobium(V) ethanolate ≥99.999% (metals basis) (max. 0.05% Ta)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Niobium(V) ethoxide

014689-09 014689-18 014689-04
AA14689-09EA 194 USD
AA14689-09 AA14689-18 AA14689-04
Niobium(V) ethanolate ≥99.999% (metals basis) (max. 0.05% Ta)
Niobium(V) ethanolate

Preparation of niobium oxide and mixed metal oxides Soluble in organic solvents. Decomposes in water


Purity: 99.999% CAS No.: 3236-82-6 Molecular Formula: C10H25NbO5 Formula Weight: 318.21 Flash Point: 36°(97°F) Physical Form: Liquid Density: 1.258 Refractive Index: 1.5160 Sensitivity: Moisture Sensitive Melting Point: 6° Boiling Point: 140-142°/0.1mm Solubility: Soluble in organic solvents. Decomposes in water Beilstein Reference: 3678883 UN No.: 2924 MDL No.: MFCD00015122


Preparation of niobium oxide and mixed metal oxides

Formula: C₁₀H₂₅NbO₅
Boiling Pt: 140…142 °C (0.1 mmHg)
Melting Pt: 6 °C
Density: 1.258
Flash Pt: 36 °C (97 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015122
CAS Number: 3236-82-6
EINECS: 221-795-2
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR