Germanium(IV) ethoxide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate
MDL: MFCD00015121
Soluble in anhydrous alcohol and benzene. Hydrolyzes in water
Purity: 97%
CAS No.: 14165-55-0
Molecular Formula: C8H20GeO4
Formula Weight: 252.84
Flash Point: 49°(121°F)
Physical Form: Liquid
Density: 1.14
Refractive Index: 1.4070
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Alcohol-like
Melting Point: -72°
Boiling Point: 185.5°
Solubility: Soluble in anhydrous alcohol and benzene. Hydrolyzes in water
UN No.: 1993
MDL No.: MFCD00015121
Formula:
C₈H₂₀GeO₄ Boiling Pt: 185,5 °C Melting Pt: –72 °C Density: 1,14 Flash Pt: 49 °C (121 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015121 CAS Number: 14165-55-0 EINECS: 238-007-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...