Calcium hydroxide ≥95.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Slaked lime
Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
Purity: 95.0% min
CAS No.: 1305-62-0
Molecular Formula: CaH2O2
Formula Weight: 74.09
Physical Form: Powder
Density: 2.24
Appearance: White
Odor: Odorless
Solubility: Slightly soluble in water. Soluble in glycerol, acids. Insoluble in alcohol
Merck Reference: 14,1673
UN No.: 3262
MDL No.: MFCD00010901
Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
Formula:
Ca(OH)₂ Melting Pt: 550 °C Density: 2.24 Storage Temperature: Ambient |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 EINECS: 215-137-3 UN: 3262 ADR: 8,III Merck Index: 14,1673 |
Specification Test Results
CaCO3 | 3.0 |
Insoluble in hydrochloric acid | 0.03% |
Cl | 0.03% |
Sulfur compounds (as SO3) | 0.1% |
Heavy Metals (as Pb) | 0.003% |
Fe | 0.05% |
MG | 0.5% |
K | 0.05% |
NA | 0.05% |
Sr | 0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...