Ammonium hydrogen difluoride ≥95%, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00012018
Purity: 95%
CAS No.: 1341-49-7
Molecular Formula: (NH4)HF2
Formula Weight: 57.04
Physical Form: Flake
Density: 1.500
pH: 2-3
Melting Point: 120-122°
Boiling Point: 238°
Merck Reference: 14,498
UN No.: 1727
MDL No.: MFCD00012018
Formula:
NH₄HF₂ Boiling Pt: 238 °C Melting Pt: 120…122 °C Density: 1.500 Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS Number: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 14,498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...