Antimony ≥99.999% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

014640-03 014640-14 014640-06
AA14640-03EA 35.6 USD
AA14640-03 AA14640-14 AA14640-06
Antimony ≥99.999% (metals basis), powder -200 mesh
Antimony

MDL: MFCD00134030


Purity: 99.999% CAS No.: 7440-36-0 Odor: Odorless UN No.: 2871 MDL No.: MFCD00134030

Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR