Mercury(II) nitrate monohydrate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate
014497-18
014497-30
AA14497-18EA
93.87
USD
AA14497-18
AA14497-30
Mercury(II) nitrate monohydrate ≥98.0% ACS
Mercury (II) nitrate monohydrate
MDL: MFCD00149736
Purity: 98.0% min
CAS No.: 7783-34-8
Molecular Formula: HgN2O6.xH2O (x=1-2)
Formula Weight: 324.60(anhy)
Physical Form: Crystalline
Density: 4.39
Appearance: White
Melting Point: 79°
Merck Reference: 14,5880
UN No.: 1625
MDL No.: MFCD00149736
Formula:
Hg(NO₃)₂·1H₂O MW: 342.62 g/mol Melting Pt: 79 Density: 4.39 Storage Temperature: Ambient |
MDL Number:
MFCD00149736 CAS Number: 7783-34-8 EINECS: 233-152-3 UN: 1625 ADR: 6.1 II Merck Index: 14,5880 |
Specification Test Results
Residue after reduction | 0.01% |
Cl | 0.002% |
SO3 | 0.002% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...