Cesium nitrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitric acid cesium salt

014440-22 014440-06 014440-14 014440-36
AA14440-22EA 289.52 USD
AA14440-22 AA14440-06 AA14440-14 AA14440-36
Cesium nitrate ≥99.99% (metals basis)
Cesium nitrate

MDL: MFCD00010963


Purity: 99.99% CAS No.: 7789-18-6 Molecular Formula: CsNO3 Formula Weight: 194.91 Physical Form: Crystalline Density: 3.68 Refractive Index: 1.55 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 414° Boiling Point: dec. Merck Reference: 14,2015 UN No.: 1451 MDL No.: MFCD00010963

Formula: CsNO₃
Boiling Pt: Dec.
Melting Pt: 414 °C
Density: 3.68
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1,III
Merck Index: 14,2015

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR