Specpure® Sulfur, Plasma Standard Solution, S 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014436-AE
014436-AP
014436-AD
AA14436-AEEA
122.37
USD
AA14436-AE
AA14436-AP
AA14436-AD
Specpure® Sulfur, Plasma Standard Solution, S 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Sulfur standards
Molecular Formula: (NH4)2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00085316
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...