Barium carbonate ≥99.8%
Supplier: Thermo Scientific Chemicals
|
Warning
|
014341-A1
014341-A7
014341-18
014341-30
AA14341-A1EA
123
USD
AA14341-A1
AA14341-A7
AA14341-18
AA14341-30
Barium carbonate ≥99.8%
Barium carbonate
MDL: MFCD00003448
Notes: m.p. 174°C (90 atm), 811°C (1 atm)
Purity: 99.8%
CAS No.: 513-77-9
Molecular Formula: BaCO3
Formula Weight: 197.35
Physical Form: 1 Micron Powder
Density: 4.43
Melting Point: 811°
Boiling Point: dec.
Merck Reference: 14,969
MDL No.: MFCD00003448
Formula:
BaCO₃ Boiling Pt: Dec. Melting Pt: 811 °C Density: 4.43 Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III Merck Index: 14,969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...