Potassium thiocyanate ≥98.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium rhodanide
MDL: MFCD00011413
Beilstein Registry No.: 3594799
Soluble in water, acetone, and alcohol. When dissolved in its own weight of water, the temperature drops ≈30°C
Fieser: 1,954
Purity: 98.5% min
CAS No.: 333-20-0
Molecular Formula: KSCN
Formula Weight: 97.18
Physical Form: Crystalline
Density: 1.89
pH: 5.3-8.5
Melting Point: 172-175°
Boiling Point: 500° dec.
Solubility: Soluble in water, acetone, and alcohol. When dissolved in its own weight of water, the temperature drops ≈30°
Beilstein Reference: 3594799
Merck Reference: 14,7691
MDL No.: MFCD00011413
Formula:
KSCN Boiling Pt: 500 °C (dec.) Melting Pt: 172…175 °C Density: 1.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 14,7691 |
Specification Test Results
Insoluble in water | 0.005% |
pH of a 5 % solution | 5.3-8.7 at 25° |
Cl | 0.005% |
SO3 | 0.005% |
NH3 | 0.003% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 2 ppm |
NA | 0.005% |
Iodine-consuming substances | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...