Sodium chlorate ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
014267-22
014267-A1
014267-D9
AA14267-22EA
45.01
USD
AA14267-22
AA14267-A1
AA14267-D9
Sodium chlorate ≥99%
Sodium chlorate
MDL: MFCD00003479
Soluble in water
Purity: 99% min
CAS No.: 7775-09-9
Molecular Formula: NaClO3
Formula Weight: 106.45
Physical Form: Granular
Density: 2.5
Melting Point: 248°
Solubility: Soluble in water
Merck Reference: 14,8598
UN No.: 1495
MDL No.: MFCD00003479
Formula:
ClNaO₃ MW: 106.44 g/mol Boiling Pt: 300 Melting Pt: 260 Density: 2.5 Storage Temperature: Ambient |
MDL Number:
MFCD00003479 CAS Number: 7775-09-9 EINECS: 231-887-4 UN: 1495 ADR: 5.1 II REACH: 01-2119474389-23 Merck Index: 14,8598 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...