Copper(II) bromide Cu ≥ 28,1%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cupric bromide, Cuprous bromide
014258-22
014258-36
AA14258-22EA
62
USD
AA14258-22
AA14258-36
Copper(II) bromide Cu ≥ 28,1%, REacton®, Reagent Grade
Copper (II) bromide
MDL: MFCD00010970
Fieser: 1,161 2,84 5,158 6,138 14,100 15,100 18,111 19,106 21,143
CAS No.: 7789-45-9
Molecular Formula: CuBr2
Formula Weight: 223.36
Physical Form: Crystalline
Density: 4.710
Sensitivity: Hygroscopic
Appearance: Dark grey to black
Odor: Odorless
Melting Point: 498°
Boiling Point: 900°
Merck Reference: 14,2629
UN No.: 3260
MDL No.: MFCD00010970
Formula:
Br₂Cu MW: 223.35 g/mol Boiling Pt: 900 Melting Pt: 498 Density: 4.710 Storage Temperature: Ambient |
MDL Number:
MFCD00010970 CAS Number: 7789-45-9 EINECS: 232-167-2 UN: 3260 ADR: 8 III Merck Index: 14,2629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...