Iron 99+% (metals basis), wire, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

014246-H2
AA14246-H2EA 684.33 USD
AA14246-H2
Iron 99+% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Iron

MDL: MFCD00010999


Purity: 99+% CAS No.: 7439-89-6 Physical Form: ≈25g/m MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR