Iron, Oil Based Standard Solution, Specpure®, Fe 1000 µg/g, Thermo Scientific

Supplier: Thermo Scientific Chemicals

014220-18
AA14220-18EA 74.9 USD
AA14220-18
Iron, Oil Based Standard Solution, Specpure®, Fe 1000 µg/g, Thermo Scientific
Standards Cationic Standards Iron Standards

Physical Form: Liquid
MDL No.: MFCD00010999

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR