Manganese, Oil Based Standard Solution, Specpure®, Mn 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014214-18
AA14214-18EA
81.73
USD
AA14214-18
Manganese, Oil Based Standard Solution, Specpure®, Mn 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Manganese Standards
Physical Form: Liquid
MDL No.: MFCD00011111
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...