Nickel, Oil Based Standard Solution, Specpure®, Ni 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014211-18
AA14211-18EA
74.6
USD
AA14211-18
Nickel, Oil Based Standard Solution, Specpure®, Ni 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Physical Form: Liquid
MDL No.: MFCD00011137
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...