Phosphorus, Oil based standard solution, Specpure®, P 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014210-18
AA14210-18EA
73.45
USD
AA14210-18
Phosphorus, Oil based standard solution, Specpure®, P 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Phosphorus Standards
Physical Form: Liquid
MDL No.: MFCD00133771
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...